missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HTRA2/Omi Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1458-SP
This item is not returnable.
View return policy
Description
HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| HTRA2/Omi | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O43464 | |
| HTRA2 | |
| E. coli-derived recombinant human HTRA2/Omi Ala134-Glu458 Accession # O43464 | |
| 25 μg | |
| Primary | |
| Detects the mitochondria-processed form of human HTRA2/Omi. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 229926 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, Omi, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27429 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction