Learn More
Human IFN-alpha Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB93454-100
 View more versions of this product
Product Code. 18752217
  • € 466.00 / 100µg
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-alpha
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
HEK293 human embryonic kidney cell line transfected with human IFN-alpha 2, Met1-Glu188, Accession # P01563
100 μg
3440
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG1
Immunocytochemistry
995514
Immunocytochemistry 8-25 μg/mL
P01563
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human IFN-alpha 2a,4a,6,7,8,10,16, and 17 in direct ELISAs.
Human
Supernatant