missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha I Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 193.00 - € 423.00
Specifications
| Antigen | IFN-alpha I/IFNA17 |
|---|---|
| Clone | 995842 |
| Dilution | Neutralization 1.5-15 ng/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762327
![]() |
R&D Systems
MAB10100-SP |
25 μg |
€ 193.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722897
![]() |
R&D Systems
MAB10100-100 |
100 μg |
€ 423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha I/IFNA17 Monoclonal specifically detects IFN-alpha I/IFNA17 in Human samples. It is validated for Neutralization.Specifications
| IFN-alpha I/IFNA17 | |
| Neutralization 1.5-15 ng/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P01571 | |
| 3451 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 995842 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNA, IFNA17, IFNalpha I, IFN-alpha-17, IFN-alphaI, INFA, interferon alpha-17, Interferon alpha-88, Interferon alpha-I', Interferon alpha-T, interferon alpha-WA, interferon, alpha 17, LeIF I, LEIF2C1 | |
| Human embryonic kidney cell line HEK293-derived recombinant human IFN-alpha I, Met1-Asp189, Accession # P01571 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-alpha I, -alpha 2, -4 alpha , -5, -6, -7, -10, -16, -17,and -21 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title