missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 214.00 - € 425.00
Specifications
| Antigen | IFN-beta |
|---|---|
| Clone | 937912 |
| Dilution | Neutralization 0.2-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Neutralization, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745479
![]() |
R&D Systems
MAB8141-SP |
25 μg |
€ 214.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713609
![]() |
R&D Systems
MAB8141-100 |
100 μg |
€ 425.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization, Immunocytochemistry.Specifications
| IFN-beta | |
| Neutralization 0.2-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-beta in direct ELISAs. |
| 937912 | |
| Neutralization, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 3456 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title