missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-gamma Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2851U-100UG
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IFN-gamma | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| E. coli-derived recombinant human IFN-gamma , Gln24-Gln166, Accession No. AAP20098.1 | |
| 100 μg | |
| 3458.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 25718 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| AAP20098.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IFN-gamma . | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction