missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-gamma DuoSet ELISA

Product Code. 18794767
Change view
Click to view available options
Sufficient For:
15 x 96-well Plates
5 x 96-well Plates
Unit Size:
1 set
Pack of 15
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18794767 15 x 96-well Plates Pack of 15
30021477 15 x 96-well Plates 1 set
30000522 5 x 96-well Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18794767 Supplier R&D Systems™ Supplier No. DY285B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human IFN-gamma DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IFN-gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IFN-gamma.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 9.38 - 600pg/mL
Immunoassay Kit Format 96-well Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sufficient For 15 x 96-well Plates
Target Species Human
Gene ID (Entrez) 3458 (Human); 15978 (Mouse); 25712 (Rat); 396991 (Porcine); 281237 (Bovine); 403801 (Canine); 493965 (Feline)
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target Interferon gamma

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.