missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-gamma Quantikine ELISA Kit

Product Code. 18751667
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18751667

Brand: R&D Systems™ SIF50C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

solid phase ELISA designed to measure human IFN-gamma levels in cell culture supernates, serum, and plasma.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 to 1000 pg/mL
Assay Sensitivity 5.69 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum
Specificity Natural and Recombinant Human IFN-gamma
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3458
Quantity 1 Pk. (6 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Target IFN-gamma
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.