missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgA Monoclonal Antibody (3-F5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550628
This item is not returnable.
View return policy
Description
Positive control: Human serum. Predicted band size: 37 kDa Subcellular Location: Cell membrane, Immunoglobulin, Membrane, Secreted.
Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
Specifications
| Human IgA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P01876, P01877 | |
| IGHA1, IGHA2 | |
| Native protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 3-F5 | |
| Unconjugated | |
| IGHA1 | |
| A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker) | |
| Mouse | |
| Protein A | |
| RUO | |
| 3493, 3494 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction