missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgA2 Rabbit anti-Human, Clone: RM125 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14953.50ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against native human IgA2.
- High specificity
- High reactivity
Specifications
| Human IgA2 | |
| Monoclonal | |
| Biotin | |
| ELISA (0.05-0.2 ug/well for capture, 0.05-0.2 ug/mL for detection) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-1 ug/mL) The optimal working dilution should be deter | |
| P01877 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3494 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| RM125 | |
| Rabbit monoclonal antibody raised against native human IgA2. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| IGHA2 | |
| Native purified human IgA2. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction