missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgD Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9857R-100UG
This item is not returnable.
View return policy
Description
IgD Monoclonal specifically detects IgD in Human samples. It is validated for Flow Cytometry.
Specifications
| IgD | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IgD in Flow Cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| IA6-2 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Human IgD, | |
| 100 μg | |
| 3495 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction