missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgE Rabbit anti-Human, Clone: RM122 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14955.50ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against native human IgE.
- High specificity
- High reactivity
Specifications
| Human IgE | |
| Monoclonal | |
| Biotin | |
| ELISA (0.01-0.1 ug/well for capture, 0.01-0.1 ug/mL for detection) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01854 | |
| IGHE | |
| Native purified human IgE. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
| RM122 | |
| Rabbit monoclonal antibody raised against native human IgE. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| IgE | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3497 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction