missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-rP10 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5137-SP
This item is not returnable.
View return policy
Description
IGFBP-rP10/KAZALD1 Polyclonal specifically detects IGFBP-rP10/KAZALD1 in Human samples. It is validated for Western Blot.
Specifications
| IGFBP-rP10/KAZALD1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bone- and odontoblast-expressed gene 1, BONO1, FKSG28, FKSG40, FLJ24094, IGFBP-related protein 10, IGFBPrP10, IGFBP-rP10, KAZALD1, Kazal-type serine peptidase inhibitor domain 1, Kazal-type serine protease inhibitor domain 1, kazal-type serine protease inhibitor domain-containing protein 1, novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein, UNQ2945/PRO21184 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 81621 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q96I82 | |
| KAZALD1 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-rP10 isoform 1 Arg31-Tyr304 Accession # Q96I82 | |
| 25 μg | |
| Primary | |
| Detects human IGFBP-rP10 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse IGFBP-rP10 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction