missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems F0135
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
Specifications
| IgG | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| Immunoglobulin G, ImmunoglobulinG | |
| Human IgG | |
| 100 Tests | |
| 3500 | |
| Human | |
| IgG |
| Flow Cytometry | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Goat | |
| Affinity Purified | |
| Secondary | |
| This antibody reacts with heavy chains on human IgG and with light chains on all human immunoglobulins. No reactivity is observed to non-immunoglobulin human, mouse, rabbit or bovine serum proteins by IEP. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction