missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TL1A/TNFSF15 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB74422G-100UG
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Flow Cytometry.
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1 | |
| E. coli-derived recombinant human TL1A/TNFSF15, Leu72-Leu251, Accession No. O95150 | |
| 100 μg | |
| 9966 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2116A | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O95150 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human TL1A/TNFSF15 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction