missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG (gamma heavy chain) Rabbit anti-Human, Clone: RM116 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14948.50ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against native human IgG (gamma heavy chain).
- High specificity
- High reactivity
Specifications
| Human IgG (gamma heavy chain) | |
| Monoclonal | |
| Biotin | |
| ELISA (0.05-0.2 ug/well for capture, 0.05-0.2 ug/mL for detection) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) The optimal working dilution should be deter | |
| P01857;P01859;P01860;P01861 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3500/3501/3502/3503 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| RM116 | |
| Rabbit monoclonal antibody raised against native human IgG (gamma heavy chain). | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| IGHG1 | |
| Native purified human IgG. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction