Learn More
Human IgG Kappa chain Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67761-1-IG-20UL
This item is not returnable.
View return policy
Description
The Human IgG Kappa chain antibody from Proteintech is a mouse monoclonal antibody generated with nature protein of Human IgG Kappa chain. This antibody recognizes human antigen. The Human IgG Kappa chain antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| IgG Kappa chain | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| Human | |
| ∼25 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 4D11F7 | |
| Unconjugated | |
| nature protein | |
| 20 μL | |
| Primary | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |