missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG Recombinant Mouse Monoclonal Antibody (17F12)
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551867
This item is not returnable.
View return policy
Description
This antibody binds to all four human IgG subtypes (IgG1, IgG2, IgG3 & IgG4). Some cross-reactivity with rabbit IgG, cynomolgus monkey IgG1 and rhesus macaque IgG1 has also been reported.
Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Specifications
| Human IgG | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Mouse | |
| Protein A | |
| RUO | |
| Human, Rhesus Macaque, Cynomolgus Monkey | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 17F12 | |
| Unconjugated | |
| Immunoglobulin G; ImmunoglobulinG | |
| This antibody was raised against human IgG. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction