missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgM Recombinant Isotype Control (SPE-7)
Description
Human IgM Isotype Control for Flow
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications
Specifications
| Antigen | Human IgM |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | SPE-7 |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH |
| Host Species | Human |
| Purification Method | Protein A/G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?