missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Membrane Form Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2001N-100UG
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-1 alpha/IL-1F1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P01583 | |
| IL1A | |
| E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583 | |
| RUO | |
| 3552 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 3405 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-1 alpha/IL-1F1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-1 alpha , recombinant rat IL-1 alpha , recombinant cotton rat IL-1 alpha , recombinant human (rh) IL-1 beta or rhIL-18 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction