missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 RAcP/IL-1 R3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB676-SP
This item is not returnable.
View return policy
Description
IL-1 RAcP/IL-1 R3 Monoclonal specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| IL-1 RAcP/IL-1 R3 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9NPH3 | |
| IL1RAP | |
| S. frugiperda insect ovarian cell line Sf21-derived recombinant human IL-1RAcP/IL-1 R3 Ser21-Glu359 (predicted) Accession # Q9NPH3 | |
| 25 μg | |
| Primary | |
| Detects human IL-1RAcP/IL-1 R3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhST2, rhIL-18 R alpha , IL-1 Rrp2, rhIL-18 RAP, rhSIGIRR, rhIL-1 RAPL1, rhIL-1 RAPL2, rhTLR1, rhTLR2, rhTLR3, rhTLR4, rhTLR5, rhTLR7, rhTLR8, rhTLR9, rhTLR10, rhMD-1, rhMD-2, or recombinant mouse RP105 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 89412 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 R3, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, IL1RAcP, IL-1RAcP, IL1RAP, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3556 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction