missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems BAF219
This item is not returnable.
View return policy
Description
IL-12 Polyclonal specifically detects IL-12 in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair).
Specifications
| IL-12 | |
| Polyclonal | |
| Biotin | |
| P29459 (p35) & P29460 (p40) | |
| IL12A | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 heterodimer Arg23-Ser219 of p35; Ile23-Ser328 of p40 Accession # P29459 (p35) & P29460 (p40) | |
| 100 μg | |
| Primary | |
| Detects human IL-12 in ELISAs and Western blots. When used in combination with the monoclonal capture antibody anti-human IL-12 p40 (Catalog # MAB609) in sandwich ELISAs, less than 2.5% cross-reactivity was observed with rhIL-12 p70. When used in combination with the monoclonal capture antibody anti-human IL-12 p70 (Catalog # MAB611) in sandwich ELISAs, less than 0.1% cross-reactivity was observed with rhIL-12 p40, rhIL-12 p35 and rmIL-12. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3592 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction