missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-12/IL-23 p40 Human ELISA Kit

For the quantitative detection of human IL-12/IL-23 p40.

€ 634.00 - € 4325.00

Specifications

Accession Number P29460,Q9NPF7
Assay Range 31.3-2,000 pg/mL
Assay Sensitivity <10 pg/mL
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human IL-12/IL-23 p40
Biotin-Conjugate anti-human IL-12/IL-23 p40 monoclonal antibody
Streptavidin-HRP
Human IL-12/IL-23 p40 Standard lyophilized, 4ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15521487
View Documents Promotion Details
Invitrogen™
BMS2013
96 Tests
€ 634.00
96 tests
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Add to basket
 
15541487
View Documents Promotion Details
Invitrogen™
BMS2013TEN
10 x 96 Tests
€ 4325.00
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 10-04-2024
to see stock.
Add to basket
 
Description

Description

The Human Interleukin-12/Interleukin-23 p40 subunit (Hu IL-12/IL-23 p40) ELISA quantitates Hu IL-12/IL-23 p40 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-12/IL-23 p40. Principle of the method The Human IL-12/IL-23 p40 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

P29460,Q9NPF7
<10 pg/mL
ELISA Kit
Human
Colorimetric Microplate Reader
CLMF,CLMF2,IL-12B,IMD28,IMD29,NKSF,NKSF2,IL-23,IL-23A,IL23P19,P19,SGRF
4.7%
HRP
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Interleukin-12 subunit beta precursor
1 hr. 20 min.
31.3-2,000 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
3593, 51561
6.4%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
Human
4 hr. 10 min.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only