Learn More
Human IL-12/IL-35 p35 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems IC15701V-100UG
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-12/IL-35 p35 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2038C | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P29460 (p40) & P29459 (P35) | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40 and human IL-12 p35 Ile23-Ser328 of p40, Arg23-Ser219 of p35 Accession # P29460 (p40) & P29459 (P35) | |
| 100 μg | |
| Detects human IL-12/IL-35 p35 in direct ELISAs. Detects the IL-12/IL-35 p35 subunit either as part of the IL-12 p40/p35 heterodimer or as the IL-12/IL-35 p35 monomer. This antibody does not detect the IL-12/IL-23 p40 subunit. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur