missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-13 R alpha 2 APC-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems FAB614A
This item is not returnable.
View return policy
Description
IL-13R alpha 2 Polyclonal specifically detects IL-13R alpha 2 in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-13R alpha 2 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL13R alpha 2, IL-13R subunit alpha-2, IL13RA2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3598 | |
| Human | |
| IgG |
| Flow Cytometry | |
| APC | |
| Q14627 | |
| IL13RA2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 2 Cys22-Leu342 Accession # Q14627 | |
| 100 Tests | |
| Primary | |
| Detects human IL-13 R alpha 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse IL-13 R alpha 2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction