missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-15 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2471U-100UG
This item is not returnable.
View return policy
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-15 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P40933 | |
| IL15 | |
| E. coli-derived recombinant human IL-15 Asn49-Ser162 Accession # P40933 | |
| RUO | |
| 3600 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 34559 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-15 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2, recombinant mouse IL-15, or rhIL-21 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction