missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-15, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17886508
Click to view available options
Quantity:
2 μg
250 μg
50 μg
Unit Size:
250µg
2µg
50µg
This item is not returnable. View return policy

Product Code. 17886508

Brand: Gibco™ AF2001550UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-15 is a 12.9 kDa protein consisting of 115 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40933
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 12.9 kDa
Name Human IL-15, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 50 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Common Name IL-15
Gene Symbol IL15
Biological Activity Determined by its ability to stimulate the proliferation of mouse CTLL-2 cells. The expected ED50 is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MNWVNVISDL KKIEDLIQSM HIDATLYTES DVHPSCKVTA MKCFLLELQV ISLESGDASI HDTVENLIIL ANNSLSSNGN VTESGCKECE ELEEKNIKEF LQSFVHIVQM FINTS
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of mouse CTLL-2 cells. The expected ED50 is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.