missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-15R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB147-SP
This item is not returnable.
View return policy
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Human samples. It is validated for Western Blot.
Specifications
| IL-15R alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| EAW86418 | |
| IL15RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-15 R alpha Ile31-Thr172 Accession # EAW86418 | |
| 25 μg | |
| Primary | |
| Detects human IL-15 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-15 R alpha , recombinant human (rh) IL-2 R alpha , rhIL-2 R beta , or rhIL-2 R gamma is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 151307 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3601 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction