missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17C Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1234G-100UG
This item is not returnable.
View return policy
Description
IL-17C Monoclonal specifically detects IL-17C in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-17C | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9P0M4.1 | |
| IL17C | |
| E. coli-derived recombinant human IL-17C His19-Val197 Accession # Q9P0M4.1 | |
| RUO | |
| 27189 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 177114 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-17C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-17A, B, D, E or F is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction