missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17E/IL-25 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC1258A
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64806 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 182203 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q9H293 | |
| IL25 | |
| E. coli-derived recombinant human IL-17E Tyr33-Gly177 Accession # Q9H293 | |
| 100 Tests | |
| Primary | |
| Detects human IL-17E in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-17E and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, or rhIL-17F. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction