missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-17E/IL-25 XL Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
100 tests
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | IL-17E/IL-25 |
| Gene ID (Entrez) | 64806 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction