missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17RC PE-conjugated Antibody (Clone 309822), R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB22691P
This item is not returnable.
View return policy
Description
IL-17RC Monoclonal specifically detects IL-17RC in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-17RC | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84818 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 309822 | |
| Flow Cytometry 10 uL/10^6 cells | |
| NP_116121 | |
| IL17RC | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 RC Leu21-Ala454 Accession # NP_116121 | |
| 100 Tests | |
| Primary | |
| Detects human IL-17 RC in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-17 R, rhIL-17 BR, rhIL-17 RD, or recombinant mouse IL-17 RC is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction