Learn More
Human IL-17RD/SEF Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB22751G-100UG
This item is not returnable.
View return policy
Description
IL-17RD/SEF Monoclonal specifically detects IL-17RD/SEF in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-17RD/SEF | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ35755, IL-17 RD, IL17RD, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLM, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, SEF, Sef homolog, SEFhSef | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IL-17 RD/SEF in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 309511 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q8NFM7 | |
| IL17RD | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 RD/SEF, Ala27-Arg299, Accession # Q8NFM7 | |
| 100 μg | |
| 54756 | |
| Human | |
| Supernatant |