Learn More
Human IL-17RD/SEF Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB22751G-100UG
Product Code. 18782479
  • € 527.00 / 100µg
Description
IL-17RD/SEF Monoclonal specifically detects IL-17RD/SEF in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17RD/SEF
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
FLJ35755, IL-17 RD, IL17RD, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLM, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, SEF, Sef homolog, SEFhSef
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human IL-17 RD/SEF in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1
Flow Cytometry
309511
Flow Cytometry 0.25-1 μg/106 cells
Q8NFM7
IL17RD
Mouse myeloma cell line NS0-derived recombinant human IL-17 RD/SEF, Ala27-Arg299, Accession # Q8NFM7
100 μg
54756
Human
Supernatant