missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18 BPa Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF119
This item is not returnable.
View return policy
Description
IL-18 BPa/IL18BP Polyclonal specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for Western Blot, ELISA, Neutralization.
Specifications
| IL-18 BPa/IL18BP | |
| Polyclonal | |
| Unconjugated | |
| O95998 | |
| IL18BP | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 BPa Thr31-Gly194 Accession # O95998 | |
| 100 μg | |
| Primary | |
| Detects human IL-18 BPa in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Neutralization 1-5 ug/mL | |
| IL18 BPa, IL18BP, IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10068 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction