missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18/IL-1F4 Propeptide Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC646V-100UG
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q14116 | |
| IL18 | |
| E. coli-derived recombinant human IL-18/IL-1F4 Accession # Q14116 | |
| RUO | |
| 3606 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 74801 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects the pro region of human IL-18/IL-1F4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-18, recombinant mouse IL-18 or recombinant rat IL-18. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction