missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18 R beta/IL-1 R7 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF118
This item is not returnable.
View return policy
Description
IL-18 R beta/IL-1 R7/ACPL Polyclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-18 R beta/IL-1 R7/ACPL | |
| Polyclonal | |
| Unconjugated | |
| O95256 | |
| IL18RAP | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 R beta/IL-1 R7 Phe20-Arg356 Accession # O95256 | |
| 100 μg | |
| Primary | |
| Detects human IL-18 R beta in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse IL-18 R beta (AcPL), recombinant human (rh) IL-18 R, rhIL-1 R1, rhIL-1 RII, and rhIL-1 Rrp2 is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.3-1.2 ug/mL | |
| Accessory protein-like, AcPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-1 R7, IL-18 receptor accessory protein, IL-18 receptor beta, IL18R beta, IL-18RAcP, IL18RAP, IL-18Rb, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8807 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction