missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Recognizes human IL1R1
- Interleukin-1 polypeptides are pleiotropic cytokines exerting diverse functions in tissues
- IL1R1 (CD121a) is a receptor for interleukin-1 alpha, interleukin-1 beta and interleukin-1 receptor antagonist
- Receptor contains large cytoplasmic region and extracellular, IL-1 binding portion composed of three immunoglobulin-like domains
- Expression found in cells including T cells, macrophages, neutrophils, eosinophils, basophils, mast cells, fibroblasts, endothelial cells
- Can shed from cell membrane as soluble form
- Function as soluble decoy receptor and prevent binding of IL-1 alpha, IL1 beta, IL-1Ra to membrane IL-1R1
Process:
- Antibody specific for IL1R1 pre-coated onto microwells
- IL1R1 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody of biotinylated specific for IL1R1 is added to detect captured IL1R1 protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set 630 nm
Specifications
Specifications
| Accession Number | BC075063 |
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 5 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 3554 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human IL-1R1 ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?