missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC202G-100UG
This item is not returnable.
View return policy
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_000577 | |
| IL2 | |
| E. coli-derived recombinant human IL-2 Ala21-Thr153 Accession # NP_000577 | |
| RUO | |
| 3558 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 5334 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-2 in direct ELISAs. Does not cross-react with recombinant IL-2 from mouse, rat, pig, or cotton rat. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction