missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-22 DuoSet ELISA
Click to view available options
Packaging Quantity:
15 Plates
5 Plates
Unit Size:
1 set
Description
The Human IL-22 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-22.
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 50616 |
| Synonym | Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 |
| Packaging Quantity | 5 Plates |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction