missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-22 R alpha 1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2770
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for Western Blot.
Specifications
| IL-22R alpha 1 | |
| Polyclonal | |
| Biotin | |
| Q8N6P7 | |
| IL22RA1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 Accession # Q8N6P7 | |
| 50 μg | |
| Primary | |
| Detects human IL-22 R alpha 1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) IL-20 R alpha , rhIL-20 R beta , and rhIL-22BP is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 58985 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction