missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-22BP Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10871G-100UG
This item is not returnable.
View return policy
Description
IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-22BP/IL22 RA2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q969J5 | |
| IL22RA2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro263 Accession # Q969J5 | |
| 100 μg | |
| Primary | |
| Detects human IL-22BP in direct ELISAs. In direct ELISAs, no cross-reactivity with mouse IL-22BP is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 875504 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 116379 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction