missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-24 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF1965-SP
This item is not returnable.
View return policy
Description
IL-24 Polyclonal specifically detects IL-24 in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| IL-24 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C49A, FISP, IL10B, IL24, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, MOB-5, ST16, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11009 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.2-0.6 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q2YHE5 | |
| IL24 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-24 Gln50-Leu206 Accession # Q2YHE5 | |
| 25 μg | |
| Primary | |
| Detects human IL-24 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction