Learn More
Human IL-28A/IFN-lambda 2 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB1587-SP
Product Code. 18740937
  • € 208.00 / 25µg
Description
IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-28A/IFN-lambda 2
Monoclonal
LYOPH
Neutralization 0.25-1.0 ug/mL
Q8IZJ0
IFNL2
Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2 Val26-Val200 Accession # Q8IZJ0
25 μg
Primary
Detects human IL-28A/IFN-lambda 2 in direct ELISAs. In direct ELISAs, 83% cross-reactivity with recombinant human (rh) IL-28B/IFN-lambda 3 and no cross-reactivity with recombinant mouse IL-28B/IFN-lambda 3 or rhIL-29/IFN-l1 is observed.
Human
Purified
Neutralization
248526
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
interleukin-28A, IFN-lambda 2, IL28A, IL-28A, interferon, lambda 2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
282616
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.