Learn More
Human IL-28A/IFN-lambda 2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1587-SP
This item is not returnable.
View return policy
Description
IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for Neutralization.
Specifications
| IL-28A/IFN-lambda 2 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.25-1.0 ug/mL | |
| Q8IZJ0 | |
| IFNL2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2 Val26-Val200 Accession # Q8IZJ0 | |
| 25 μg | |
| Primary | |
| Detects human IL-28A/IFN-lambda 2 in direct ELISAs. In direct ELISAs, 83% cross-reactivity with recombinant human (rh) IL-28B/IFN-lambda 3 and no cross-reactivity with recombinant mouse IL-28B/IFN-lambda 3 or rhIL-29/IFN-l1 is observed. | |
| Human | |
| Purified |
| Neutralization | |
| 248526 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| interleukin-28A, IFN-lambda 2, IL28A, IL-28A, interferon, lambda 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 282616 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |