missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-31 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2824-SP
This item is not returnable.
View return policy
Description
IL-31 Polyclonal specifically detects IL-31 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-31 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL31, IL-31interleukin-31, interleukin 31 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 386653 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q6EBC2 | |
| IL31 | |
| E. coli-derived recombinant human IL-31 Ser24-Thr164 Accession # Q6EBC2 | |
| 25 μg | |
| Primary | |
| Detects human IL-31 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant mouse IL-31 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction