missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-34 Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 209.00 - € 602.00
Specifications
| Antigen | IL-34 |
|---|---|
| Clone | 578410 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18756819
![]() |
R&D Systems
MAB52651-SP |
25 μg |
€ 209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18111411
![]() |
R&D Systems
MAB52651 |
500 μg |
€ 602.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| IL-34 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C16orf77, IL34, IL-34, interleukin 34 | |
| IL34 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 578410 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6ZMJ4 | |
| 146433 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242 Accession # Q6ZMJ4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-34 in sandwich immunoassays. In this format, no cross-reactivity with recombinant mouse IL-34 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title