missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-37/IL-1F7 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB19751U-100UG
This item is not returnable.
View return policy
Description
IL-37/IL-1F7 Monoclonal specifically detects IL-37/IL-1F7 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-37/IL-1F7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9NZH6 | |
| IL37 | |
| E. coli-derived recombinant human IL-37/IL-1F7 Lys53-Val206 Accession # Q9NZH6 | |
| 100 μg | |
| Primary | |
| Detects human IL-37/IL-1F7 in ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 899826 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27178 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction