missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4 APC-conjugated Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems IC11474A-100
This item is not returnable.
View return policy
Description
IL-4 Monoclonal antibody specifically detects IL-4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| IL-4 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Chinese Hamster Ovary cell line, CHO-derived human IL-4, His25-Ser153, Accession # P05112 | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining) | |
| 1067629 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P05112 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3565 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction