missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems BAF204
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
IL-4 Polyclonal specifically detects IL-4 in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).
Tekniske data
| IL-4 | |
| Polyclonal | |
| Biotin | |
| NP_000580 | |
| IL4 | |
| E. coli-derived recombinant human IL-4 (R&D Systems, Catalog # 204-IL) His25-Ser153 Accession # NP_000580 | |
| 50 μg | |
| Primary | |
| Detects human IL-4 in ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3565 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion