missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 242.00 - € 2185.00
Specifications
| Antigen | IL-6 |
|---|---|
| Clone | 1936 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 1-25 ug/mL, Neutralization 0.05-0.15 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Immunohistochemistry, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707129
![]() |
R&D Systems
MAB2061-SP |
25 μg |
€ 242.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18159440
![]() |
R&D Systems
MAB2061-100 |
100 μg |
€ 534.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18192461
|
R&D Systems
MAB2061-500 |
500 μg |
€ 2185.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Immunohistochemistry, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| IL-6 | |
| LYOPH | |
| Immunohistochemistry, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| IL6 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1936 | |
| Immunohistochemistry 1-25 ug/mL, Neutralization 0.05-0.15 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| P05231 | |
| 3569 | |
| E. coli-derived recombinant human IL-6 Val30-Met212 Accession # P05231 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-6 in direct ELISAs. Does not cross-react with recombinant IL-6 from mouse, rat, or pig. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title