missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-6 Quantikine HS ELISA
Click to view available options
Quantity:
1 Kit
Unit Size:
96 tests
Description
The Human IL-6 Quantikine HS ELISA Kit quantifies human IL-6 in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), urine (100 ul).
Specifications
Specifications
| Assay Range | 0.16 to 10 pg/mL |
| Assay Sensitivity | 0.09 pg/mL |
| Sample Type | Plasma (EDTA, Heparin), Serum, Serum, Urine, Urine |
| Specificity | Natural and Recombinant Human IL-6 |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3569 |
| Quantity | 1 Kit |
| Sample Volume | Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Urine (100uL) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction