missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Importin alpha 2/KPNA2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6207
This item is not returnable.
View return policy
Description
Importin alpha 2/KPNA2 Polyclonal specifically detects Importin alpha 2/KPNA2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Importin alpha 2/KPNA2 | |
| Polyclonal | |
| Unconjugated | |
| P52292 | |
| KPNA2 | |
| E. coli-derived recombinant human Importin alpha 2/KPNA2 Met1-Asp132 Accession # P52292 | |
| 100 μg | |
| Primary | |
| Detects human Importin alpha 2/KPNA2 in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, KPNA2, Pendulin, QIP2, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 3838 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction